siRNA Details
| Gene Symbol: Pvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021424.2 | NP_067399.2 | 3 | 748 | ||
| GenBank mRNA | AF239762.1 | - | 3 | 680 | ||
| AF270977.1 | - | 3 | 680 | |||
| AF297665.1 | - | 3 | 695 | |||
| AK147479.1 | - | 1340 | ||||
| BC060694.1 | - | 3 | 748 | 6405088 | ||

Pvrl1
Genomic Map