siRNA Details
| Gene Symbol: Nudt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021431.2 | NP_067406.2 | 2 | 1413 | ||
| GenBank mRNA | AB041576.1 | - | 2 | 1159 | ||
| AK036077.1 | - | 2 | 744 | |||
| AK048263.1 | - | 2 | 1416 | |||
| AK053310.1 | - | 2 | 1193 | |||
| AY152853.1 | - | 2 | 1159 | |||
| BC071274.1 | - | 2 | 1217 | 6331160 | ||

Nudt11
Genomic Map