siRNA Details
| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021433.3 | NP_067408.1 | 2 | 351 | ||
| GenBank mRNA | AF237814.1 | - | 2 | 161 | ||
| AK009690.1 | - | 2 | 306 | |||
| AK018167.1 | - | 2 | 339 | |||
| AK019106.1 | - | 2 | 313 | |||
| AK031673.1 | - | 2 | 330 | |||
| AK044905.1 | - | 2 | 312 | |||
| AK049124.1 | - | 2 | 311 | |||
| AK151549.1 | - | 2 | 305 | |||
| AK159392.1 | - | 2 | 323 | |||
| BC029205.1 | - | 2 | 268 | 4039560 | ||
| BQ749205.1 | - | 2 | 324 | |||
| CJ088919.1 | - | 2 | 351 | |||

Stx6
Genomic Map