siRNA Details
| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021433.3 | NP_067408.1 | 7 | 822 | ||
| GenBank mRNA | AF237814.1 | - | 7 | 632 | ||
| AK009690.1 | - | 7 | 777 | |||
| AK019106.1 | - | 7 | 784 | |||
| AK031673.1 | - | 7 | 801 | |||
| AK044905.1 | - | 7 | 783 | |||
| AK049124.1 | - | 7 | 782 | |||
| AK151549.1 | - | 7 | 776 | |||
| AK159392.1 | - | 7 | 794 | |||
| AK163167.1 | - | 195 | ||||
| BC029205.1 | - | 7 | 739 | 4039560 | ||
| CN456515.1 | - | 3 | 202 | |||

Stx6
Genomic Map