siRNA Details
| Gene Symbol: Slc35b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021435.3 | NP_067410.1 | 8, 9 | 877 | ||
| GenBank mRNA | AB041549.1 | - | 8, 9 | 878 | ||
| AK029046.1 | - | 7, 8 | 3498 | |||
| AK029969.1 | - | 8, 9 | 935 | |||
| AK033913.1 | - | 8, 9 | 807 | |||
| AK042150.1 | - | 8, 9 | 777 | |||
| AK045791.1 | - | 8, 9 | 820 | |||
| AK138591.1 | - | 8, 9 | 818 | |||
| AK163299.1 | - | 3, 4 | 345 | |||
| BC016275.1 | - | 8, 9 | 808 | |||
| BC033512.1 | - | 8, 9 | 802 | 4017336 | ||

Slc35b4
Genomic Map