siRNA Details
| Gene Symbol: Cts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021445.1 | NP_067420.1 | 6 | 822 | ||
| GenBank mRNA | AF223401.1 | - | 6 | 822 | ||
| AK005518.1 | - | 6 | 841 | |||
| AY014781.1 | - | 6 | 838 | |||
| BC099455.1 | - | 6 | 824 | 30787134 | ||

Cts6
Genomic Map