siRNA Details
| Gene Symbol: Elp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021448.2 | NP_067423.2 | 4 | 457 | ||
| GenBank mRNA | AF291064.1 | - | 4 | 423 | ||
| AK033246.1 | - | 4 | 458 | |||
| AK034457.1 | - | 4 | 456 | |||
| AK044699.1 | - | 4 | 434 | |||
| AK050960.1 | - | 4 | 458 | |||
| AK147826.1 | - | 458 | ||||
| AK167725.1 | - | 4 | 456 | |||
| AK167853.1 | - | 4 | 456 | |||
| AK168370.1 | - | 4 | 458 | |||
| BC015284.1 | - | 4 | 449 | 4239056 | ||
| BC051943.1 | - | 4 | 449 | 5686798 | ||

Elp2
Genomic Map