siRNA Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021472.4 | NP_067447.2 | 2 | 450 | ||
| NM_201239.3 | NP_957691.1 | 2 | 747 | |||
| GenBank mRNA | AK050451.1 | - | 2 | 347 | ||
| AK076504.1 | - | 2 | 420 | |||
| AY762362.1 | - | 2 | 349 | |||
| BC061105.1 | - | 2 | 331 | 30296907 | ||

Rnase4
Genomic Map