siRNA Details
| Gene Symbol: Treh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277847.1 | NP_001264776.1 | 7 | 730 | ||
| NM_001277851.1 | NP_001264780.1 | 7 | 734 | |||
| NM_021481.3 | NP_067456.1 | 8 | 835 | |||
| GenBank mRNA | AF136944.1 | - | 8 | 830 | ||
| AK002566.1 | - | 7 | 733 | |||
| AK008912.1 | - | 8 | 820 | |||
| BC019214.1 | - | 7 | 725 | 5064708 | ||

Treh
Genomic Map