siRNA Details
| Gene Symbol: Zfp862-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028840.1 | - | 1 | 133 | ||
| AK129160.1 | - | 6 | 2699 | |||
| BC059055.1 | - | 1 | 750 | |||
| BC068148.1 | - | 6 | 2652 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp862-ps| Gene Symbol: Zfp862-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028840.1 | - | 1 | 133 | ||
| AK129160.1 | - | 6 | 2699 | |||
| BC059055.1 | - | 1 | 750 | |||
| BC068148.1 | - | 6 | 2652 | |||