siRNA Details
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037085.1 | - | 9 | 924 | ||
BC049867.1 | - | 9 | 947 | |||
BC057921.1 | - | 9 | 935 | 5101105 | ||
X99571.1 | - | 9 | 930 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037085.1 | - | 9 | 924 | ||
BC049867.1 | - | 9 | 947 | |||
BC057921.1 | - | 9 | 935 | 5101105 | ||
X99571.1 | - | 9 | 930 |