siRNA Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021498.2 | NP_067473.2 | 3 | 277 | ||
| GenBank mRNA | AF230806.1 | - | 3 | 277 | ||
| AK007693.1 | - | 472 | ||||
| BC024996.1 | - | 3 | 277 | 5253060 | ||
| BC034004.1 | - | 285 | ||||

Pole3
Genomic Map