siRNA Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021504.3 | NP_067479.2 | 12 | 1850 | ||
| GenBank mRNA | AF250927.1 | - | 12 | 1837 | ||
| AK003279.1 | - | 3 | 220 | |||
| AK028248.1 | - | 12 | 1851 | |||
| AK148931.1 | - | 12 | 1854 | |||
| AK169205.1 | - | 11 | 1673 | |||
| BC028961.1 | - | 12 | 1804 | 5366023 | ||

Ngly1
Genomic Map