siRNA Details
| Gene Symbol: Edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021519.1 | NP_067494.1 | 3 | 210 | ||
| GenBank mRNA | AB030185.1 | - | 3 | 210 | ||
| AJ309569.1 | - | 3 | 158 | |||
| AK002345.1 | - | 3 | 225 | |||
| AK003557.1 | - | 3 | 237 | |||
| AK010191.1 | - | 3 | 225 | |||
| BC023472.1 | - | 3 | 205 | 5250117 | ||

Edf1
Genomic Map