siRNA Details
| Gene Symbol: Mkks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141946.1 | NP_001135418.1 | 6 | 2227 | ||
| NM_001286981.1 | NP_001273910.1 | 7 | 2259 | |||
| NM_001286983.1 | NP_001273912.1 | 5 | 1074 | |||
| NM_021527.2 | NP_067502.2 | 6 | 2463 | |||
| GenBank mRNA | AF254074.1 | - | 6 | 2235 | ||
| AK004987.1 | - | 7 | 2016 | |||
| AK032528.1 | - | 6 | 2225 | |||
| AK032554.1 | - | 6 | 2225 | |||
| BC080765.1 | - | 6 | 2203 | |||
| BC100336.1 | - | 5 | 827 | |||
| BC117835.1 | - | 4 | 1790 | |||
| BC117836.1 | - | 4 | 1791 | 40093115 | ||

Mkks
Genomic Map