siRNA Details
| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021534.3 | NP_067509.1 | 3 | 264 | ||
| GenBank mRNA | AJ271476.1 | - | 3 | 253 | ||
| AK003127.1 | - | 3 | 279 | |||
| AK013793.1 | - | 3 | 294 | |||
| AK019381.1 | - | 3 | 279 | |||
| AK050647.1 | - | 3 | 265 | |||
| AK156743.1 | - | 264 | ||||
| BC025796.1 | - | 3 | 243 | 5100832 | ||

Pxmp4
Genomic Map