siRNA Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021537.3 | NP_067512.3 | 4 | 514 | ||
| GenBank mRNA | AF004934.1 | - | 3 | 352 | ||
| AK148041.1 | - | 4 | 488 | |||
| AK152917.1 | - | 4 | 457 | |||
| BC029875.1 | - | 4 | 416 | |||
| BC052913.1 | - | 4 | 449 | 30057101 | ||
| BC071218.1 | - | 4 | 509 | 4012695 | ||

Stk25
Genomic Map