siRNA Details
| Gene Symbol: Rnf130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290749.1 | NP_001277678.1 | 5 | 897 | ||
| NM_001290750.1 | NP_001277679.1 | 5 | 897 | |||
| NM_021540.4 | NP_067515.2 | 5 | 897 | |||
| GenBank mRNA | AF171875.1 | - | 5 | 865 | ||
| AK011088.1 | - | 5 | 882 | |||
| AK166468.1 | - | 5 | 848 | |||
| AK220529.1 | - | 4 | 2433 | |||
| BC018199.1 | - | 5 | 891 | 4017435 | ||
| BC048901.1 | - | 5 | 880 | 6390079 | ||

Rnf130
Genomic Map