siRNA Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB036882.1 | - | 8 | 1498 | ||
| AK154535.1 | - | 7 | 1439 | |||
| AK164324.1 | - | 7 | 1387 | |||
| AK164651.1 | - | 7 | 1386 | |||
| AK178833.1 | - | 1 | 31 | |||
| BC034719.1 | - | 8 | 1468 | 3710931 | ||
| BM944891.1 | - | 4 | 571 | |||

Midn
Genomic Map