siRNA Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021605.3 | NP_067618.1 | 3 | 293 | ||
| GenBank mRNA | AF217650.1 | - | 2 | 144 | ||
| AK035502.1 | - | 3 | 292 | |||
| AK078639.1 | - | 3 | 302 | |||
| AK088173.1 | - | 3 | 342 | |||
| AK149308.1 | - | 3 | 341 | |||
| AK165583.1 | - | 3 | 271 | |||
| BC026541.1 | - | 3 | 285 | |||
| BC037697.1 | - | 3 | 270 | 3994871 | ||

Nek7
Genomic Map