siRNA Details
| Gene Symbol: Ackr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276719.1 | NP_001263648.1 | 4 | 562 | ||
| NM_021609.4 | NP_067622.2 | 3 | 450 | |||
| GenBank mRNA | AK028281.1 | - | 3 | 342 | ||
| AK145960.1 | - | 4 | 447 | |||
| AK147166.1 | - | 4 | 454 | |||
| AK157140.1 | - | 4 | 563 | |||
| BC113146.1 | - | 2 | 183 | 40055448 | ||
| Y12879.1 | - | 2 | 253 | |||

Ackr2
Genomic Map