siRNA Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021713.2 | NP_068359.1 | 3, 4 | 513 | ||
| GenBank mRNA | AF289484.1 | - | 3, 4 | 505 | ||
| AK002656.1 | - | 3, 4 | 488 | |||
| AK004104.1 | - | 3, 4 | 488 | |||
| AK013232.1 | - | 3, 4 | 459 | |||
| BC023124.1 | - | 2, 3 | 640 | |||
| BC028501.1 | - | 3, 4 | 498 | 5256915 | ||

Myg1
Genomic Map