siRNA Details
| Gene Symbol: Ms4a4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021718.2 | NP_068364.1 | 3 | 280 | ||
| GenBank mRNA | AF230640.1 | - | 3 | 237 | ||
| AF237913.1 | - | 3 | 249 | |||
| AF354929.1 | - | 3 | 237 | |||
| AK039824.1 | - | 3 | 255 | |||
| AK080934.1 | - | 3 | 281 | |||
| AK088631.1 | - | 3 | 236 | |||
| AK088780.1 | - | 3 | 286 | |||
| AK138737.1 | - | 3 | 253 | |||
| AK172495.1 | - | 3 | 253 | |||
| BC028433.1 | - | 3 | 237 | 1248222 | ||

Ms4a4b
Genomic Map