siRNA Details
| Gene Symbol: Sap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021788.2 | NP_068560.1 | 2 | 645 | ||
| GenBank mRNA | AF075136.1 | - | 2 | 656 | ||
| AK010928.1 | - | 2 | 646 | |||
| AK088745.1 | - | 2 | 650 | |||
| BC132081.1 | - | 2 | 628 | 40130358 | ||
| BC132087.1 | - | 2 | 628 | 40130364 | ||

Sap30
Genomic Map