siRNA Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021790.2 | NP_068562.1 | 8 | 812 | ||
| NM_181061.5 | NP_851406.1 | 8 | 935 | |||
| GenBank mRNA | AB043687.1 | - | 8 | 812 | ||
| AK082939.1 | - | 8 | 592 | |||
| AK135773.1 | - | 328 | ||||
| AK160776.1 | - | 8 | 583 | |||
| AK165268.1 | - | 2 | 3311 | |||
| BC028655.1 | - | 8 | 948 | |||

Cenpk
Genomic Map