siRNA Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021790.2 | NP_068562.1 | 7 | 716 | ||
| NM_181061.5 | NP_851406.1 | 7 | 839 | |||
| GenBank mRNA | AB043687.1 | - | 7 | 716 | ||
| AK082939.1 | - | 7 | 496 | |||
| AK135773.1 | - | 232 | ||||
| AK160776.1 | - | 7 | 487 | |||
| AK165268.1 | - | 2 | 2923 | |||
| BC028655.1 | - | 7 | 852 | |||

Cenpk
Genomic Map