siRNA Details
| Gene Symbol: Iigp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146275.1 | NP_001139747.1 | 3 | 755 | ||
| NM_021792.4 | NP_068564.4 | 2 | 572 | |||
| GenBank mRNA | AF194871.1 | - | 1 | 467 | ||
| AJ007971.1 | - | 2 | 554 | |||
| AK089702.1 | - | 2 | 583 | |||
| AK153060.1 | - | 2 | 574 | |||
| BC004649.1 | - | 2 | 549 | 3483102 | ||
| CT010300.1 | - | 1 | 450 | |||

Iigp1
Genomic Map