siRNA Details
| Gene Symbol: Syt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021889.4 | NP_068689.2 | 2, 3 | 717 | ||
| GenBank mRNA | AB026802.1 | - | 2, 3 | 516 | ||
| AK089115.1 | - | 2, 3 | 718 | |||
| AK156631.1 | - | 2, 3 | 718 | |||
| BC132495.1 | - | 2, 3 | 704 | 40130772 | ||
| BC137904.1 | - | 2, 3 | 704 | 8860923 | ||
| BC145024.1 | - | 2, 3 | 704 | |||

Syt9
Genomic Map