siRNA Details
| Gene Symbol: Wtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113532.1 | NP_001107004.1 | 4, 5 | 345 | ||
| NM_001113533.1 | NP_001107005.1 | 4, 5 | 345 | |||
| NM_175394.2 | NP_780603.1 | 4, 5 | 393 | |||
| GenBank mRNA | AJ276707.1 | - | 2, 3 | 103 | ||
| AK031965.1 | - | 4, 5 | 401 | |||
| AK076111.1 | - | 4, 5 | 387 | |||
| AK156334.1 | - | 4, 5 | 241 | |||
| BC046416.1 | - | 4, 5 | 274 | |||
| BC047338.1 | - | 4, 5 | 274 | |||
| BC093504.1 | - | 4, 5 | 278 | |||
| BY016042.1 | - | 4, 5 | 345 | |||

Wtap
Genomic Map