siRNA Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021895.2 | NP_068695.1 | 16 | 2040 | ||
| GenBank mRNA | AJ289242.2 | - | 16 | 2040 | ||
| AK030237.1 | - | 9 | 1210 | |||
| AK145322.1 | - | 16 | 2093 | |||
| AK150044.1 | - | 16 | 2042 | |||
| BC013616.1 | - | 16 | 2049 | 3157024 | ||
| BC027032.1 | - | 16 | 1989 | |||
| BC087554.1 | - | 16 | 1993 | 5351470 | ||
| DQ303410.1 | - | 16 | 1940 | |||
| DQ303411.1 | - | 16 | 1943 | |||

Actn4
Genomic Map