siRNA Details
| Gene Symbol: Trp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021897.3 | NP_068697.1 | 3 | 715 | ||
| GenBank mRNA | AJ131776.1 | - | 3 | 716 | ||
| AK142321.1 | - | 3 | 711 | |||
| AY034612.1 | - | 3 | 716 | |||
| BC132124.1 | - | 2 | 617 | 40130401 | ||
| BC138711.1 | - | 2 | 617 | 8861733 | ||

Trp53inp1
Genomic Map