siRNA Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038699.2 | NP_001033788.1 | 3 | 370 | ||
| NM_022014.4 | NP_071297.1 | 3 | 370 | |||
| GenBank mRNA | AA058133.1 | - | 3 | 326 | ||
| AJ404616.1 | - | 3 | 356 | |||
| AK010151.1 | - | 3 | 364 | |||
| AK030362.1 | - | 3 | 357 | |||
| AK172235.1 | - | 3 | 355 | |||
| BC054374.1 | - | 3 | 351 | 6313660 | ||
| BC064710.1 | - | 3 | 356 | 30380196 | ||

Fn3k
Genomic Map