siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022018.3 | NP_071301.2 | 13, 14 | 1820 | ||
| GenBank mRNA | AB049355.1 | - | 13, 14 | 1660 | ||
| AK029847.1 | - | 9, 10 | 1132 | |||
| AK136597.1 | - | 13, 14 | 1730 | |||
| AK170193.1 | - | 14, 15 | 1898 | |||
| AK170347.1 | - | 4, 5 | 415 | |||
| BC021332.1 | - | 13, 14 | 1708 | |||
| BC038006.1 | - | 13, 14 | 1676 | |||
| BC058234.1 | - | 11, 12 | 1420 | 6306849 | ||
| BC099690.1 | - | 13, 14 | 1727 | |||
| BC137843.1 | - | 9, 10 | 1197 | 8860862 | ||

Fam129a
Genomic Map