siRNA Details
| Gene Symbol: Cables1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146287.1 | NP_001139759.1 | 1 | 1139 | ||
| NM_022021.2 | NP_071304.2 | 1 | 1139 | |||
| GenBank mRNA | AF133208.1 | - | 1 | 1138 | ||
| AF328140.1 | - | 1 | 824 | |||
| AK156273.1 | - | 1 | 1195 | |||
| AK158396.1 | - | 1 | 869 | |||
| BC145142.1 | - | 1 | 1123 | 9053693 | ||
| BC150714.1 | - | 1 | 1123 | 9087625 | ||
| CX213994.1 | - | 1 | 200 | |||

Cables1
Genomic Map