siRNA Details
| Gene Symbol: Yeats4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026570.4 | NP_080846.1 | 6, 7 | 707 | ||
| GenBank mRNA | AK002839.1 | - | 5, 6 | 355 | ||
| AK010522.1 | - | 6, 7 | 708 | |||
| AK012096.1 | - | 6, 7 | 674 | |||
| AK016290.1 | - | 1, 2 | 143 | |||
| AK081867.1 | - | 6, 7 | 718 | |||
| BC020043.1 | - | 6, 7 | 675 | 4013548 | ||

Yeats4
Genomic Map