siRNA Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146217.1 | NP_001139689.1 | 7 | 921 | ||
| NM_022316.2 | NP_071711.2 | 7 | 888 | |||
| GenBank mRNA | AF070470.1 | - | 7 | 864 | ||
| AK040931.1 | - | 7 | 922 | |||
| BC031804.1 | - | 7 | 868 | 3673732 | ||

Smoc1
Genomic Map