siRNA Details
| Gene Symbol: Popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF204175.1 | - | 3 | 1147 | ||
| AK084713.1 | - | 3 | 1179 | |||
| BC064005.1 | - | 3 | 1183 | 30287674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Popdc2| Gene Symbol: Popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF204175.1 | - | 3 | 1147 | ||
| AK084713.1 | - | 3 | 1179 | |||
| BC064005.1 | - | 3 | 1183 | 30287674 | ||