siRNA Details
| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022326.3 | NP_071721.2 | 5 | 557 | ||
| GenBank mRNA | AF202528.1 | - | 5 | 600 | ||
| AK005428.1 | - | 5 | 557 | |||
| AK005550.1 | - | 5 | 552 | |||
| AY014777.1 | - | 5 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsm| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022326.3 | NP_071721.2 | 5 | 557 | ||
| GenBank mRNA | AF202528.1 | - | 5 | 600 | ||
| AK005428.1 | - | 5 | 557 | |||
| AK005550.1 | - | 5 | 552 | |||
| AY014777.1 | - | 5 | 522 | |||