siRNA Details
| Gene Symbol: Mllt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022328.2 | NP_071723.1 | 6 | 995 | ||
| GenBank mRNA | AF298887.1 | - | 5 | 881 | ||
| AF312858.1 | - | 6 | 892 | |||
| AK144605.1 | - | 6 | 1023 | |||
| AK147804.1 | - | 3 | 599 | |||
| AK156820.1 | - | 6 | 1022 | |||
| AK180836.1 | - | 1 | 229 | |||
| AK211968.1 | - | 1 | 229 | |||
| AK212904.1 | - | 1 | 216 | |||
| BC053005.1 | - | 6 | 995 | 5716396 | ||

Mllt1
Genomic Map