siRNA Details
| Gene Symbol: Mllt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022328.2 | NP_071723.1 | 10, 11 | 1559 | ||
| GenBank mRNA | AF298887.1 | - | 9, 10 | 1445 | ||
| AF312858.1 | - | 10, 11 | 1456 | |||
| AK144605.1 | - | 10, 11 | 1587 | |||
| AK147804.1 | - | 7, 8 | 1163 | |||
| AK156820.1 | - | 10, 11 | 1586 | |||
| BC053005.1 | - | 10, 11 | 1559 | 5716396 | ||
| BC139810.1 | - | 1, 2 | 67 | |||

Mllt1
Genomic Map