siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109753.1 | NP_001103223.1 | 9 | 1829 | ||
| NM_153579.4 | NP_705807.2 | 9 | 2036 | |||
| GenBank mRNA | AK029736.1 | - | 10 | 1861 | ||
| AK043830.1 | - | 9 | 1756 | |||
| AK122359.1 | - | 8 | 981 | |||
| AK139478.1 | - | 9 | 1731 | |||
| AK140104.1 | - | 9 | 1749 | |||
| AK147261.1 | - | 9 | 1823 | |||
| AK159028.1 | - | 9 | 1734 | |||
| BC060224.1 | - | 8 | 1420 | 30356990 | ||
| BC079568.1 | - | 9 | 2036 | |||

Sv2b
Genomic Map