siRNA Details
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109753.1 | NP_001103223.1 | 9 | 1888 | ||
NM_153579.4 | NP_705807.2 | 9 | 2095 | |||
GenBank mRNA | AK029736.1 | - | 10 | 1920 | ||
AK043830.1 | - | 9 | 1815 | |||
AK122359.1 | - | 8 | 1040 | |||
AK139478.1 | - | 9 | 1790 | |||
AK140104.1 | - | 9 | 1808 | |||
AK147261.1 | - | 9 | 1882 | |||
AK159028.1 | - | 9 | 1793 | |||
BC060224.1 | - | 8 | 1479 | 30356990 | ||
BC079568.1 | - | 9 | 2095 |
