siRNA Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022413.4 | NP_071858.3 | 15 | 2353 | ||
| GenBank mRNA | AB037373.1 | - | 15 | 2260 | ||
| AJ542487.1 | - | 15 | 2058 | |||
| AK077234.1 | - | 15 | 2283 | |||
| AK168760.1 | - | 15 | 2351 | |||
| BC016101.1 | - | 15 | 2291 | 4911355 | ||

Trpv6
Genomic Map