siRNA Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022331.1 | NP_071726.1 | 6 | 895 | ||
| GenBank mRNA | AB034991.1 | - | 6 | 895 | ||
| AK081688.1 | - | 6 | 899 | |||
| AK159500.1 | - | 6 | 901 | |||
| AK165832.1 | - | 6 | 901 | |||
| AK172874.1 | - | 6 | 888 | |||
| BC013523.1 | - | 6 | 887 | 4236803 | ||

Herpud1
Genomic Map