siRNA Details
| Gene Symbol: Fndc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022424.5 | NP_071869.2 | 5, 6 | 909 | ||
| GenBank mRNA | AB030187.1 | - | 1, 2 | 81 | ||
| AK013203.1 | - | 5, 6 | 903 | |||
| AK018154.1 | - | 5, 6 | 905 | |||
| AK163183.1 | - | 5, 6 | 910 | |||
| BC027164.1 | - | 1, 2 | 63 | |||
| BC139278.1 | - | 5, 6 | 661 | 8862300 | ||
| BC139292.1 | - | 5, 6 | 661 | 8862314 | ||

Fndc4
Genomic Map