siRNA Details
| Gene Symbol: Ms4a4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029499.3 | NP_083775.1 | 3 | 350 | ||
| GenBank mRNA | AF237914.2 | - | 3 | 324 | ||
| AK017928.1 | - | 3 | 330 | |||
| AK151045.1 | - | 3 | 343 | |||
| AK153495.1 | - | 2 | 194 | |||
| AK157137.1 | - | 352 | ||||
| AK172650.1 | - | 3 | 322 | |||
| BC027496.1 | - | 3 | 306 | 1383416 | ||
| CT010170.1 | - | 2 | 218 | |||

Ms4a4c
Genomic Map