siRNA Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127351.1 | NP_001120823.1 | 6 | 821 | ||
| NM_001177804.1 | NP_001171275.1 | 6 | 813 | |||
| NM_022433.2 | NP_071878.2 | 6 | 866 | |||
| GenBank mRNA | AF299338.1 | - | 6 | 866 | ||
| AF299339.1 | - | 6 | 821 | |||
| AK075861.1 | - | 6 | 795 | |||
| BC025878.1 | - | 6 | 765 | 5253854 | ||
| CT010402.1 | - | 5 | 572 | |||
| EU886466.1 | - | 6 | 811 | |||
| FJ621493.1 | - | 6 | 809 | |||

Sirt3
Genomic Map