siRNA Details
| Gene Symbol: Sp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022435.2 | NP_071880.1 | 2 | 1475 | ||
| GenBank mRNA | AF279479.1 | - | 2 | 1232 | ||
| AK036234.1 | - | 2 | 1475 | |||
| BC116691.1 | - | 2 | 1326 | 40106236 | ||
| BC117505.1 | - | 2 | 1326 | 40106237 | ||

Sp5
Genomic Map