siRNA Details
| Gene Symbol: Inpp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290437.1 | NP_001277366.1 | 2 | 782 | ||
| NM_033134.3 | NP_149125.1 | 1 | 1066 | |||
| GenBank mRNA | AF226683.1 | - | 1 | 1028 | ||
| AK047245.1 | - | 1 | 1051 | |||
| AK080075.1 | - | 1 | 1048 | |||
| AK085439.1 | - | 2 | 785 | |||
| AK154097.1 | - | 1 | 1067 | |||
| AK158135.1 | - | 1 | 1051 | |||
| AK170174.1 | - | 1 | 999 | |||
| AK170786.1 | - | 1 | 1023 | |||
| AK170812.1 | - | 1 | 1037 | |||
| BC052717.1 | - | 1 | 1035 | 6837339 | ||
| BC057566.1 | - | 1 | 1031 | |||
| BC080295.1 | - | 1 | 1031 | 30354397 | ||

Inpp5e
Genomic Map