siRNA Details
| Gene Symbol: Nisch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022656.2 | NP_073147.2 | 17 | 3394 | ||
| GenBank mRNA | AF315344.1 | - | 13 | 2829 | ||
| AK122412.1 | - | 15 | 2910 | |||
| BC089329.1 | - | 12 | 2613 | |||
| BC108364.1 | - | 12 | 2589 | |||
| BC144980.1 | - | 13 | 2723 | |||

Nisch
Genomic Map